missing translation for 'onlineSavingsMsg'
Learn More

TREM2 Antibody (2B5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NBP1-07101

 View more versions of this product

Product Code. 18706303

  • £444.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TREM2 Monoclonal specifically detects TREM2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TREM2
Monoclonal
1.0 mg/mL
Western Blot 1:500-1000, Flow Cytometry 2-5ug, ELISA 1:500-1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Mouse
Protein G purified
RUO
Primary
Human, Rat
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
2B5
Unconjugated
Q9NZC2
TREM2
Recombinant human TREM2 (19-161aa) purified from E. coli (NP_061838).
0.1 mL
Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience
54209
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only