missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (CL7111), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £433.00
Specifications
| Antigen | TREM1 |
|---|---|
| Clone | CL7111 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603731
|
Novus Biologicals
NBP2-76531-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256719
|
Novus Biologicals
NBP2-76531 |
100 μL |
£433.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Knockdown Validated.Specifications
| TREM1 | |
| Monoclonal | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 54210 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: KLTEEKYELKEGQTLDVK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL7111 | |
| Unconjugated | |
| Human | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| TREM1 | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title