missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (CL7111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76531-25ul
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Knockdown Validated.
Specifications
| TREM1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| TREM1 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: KLTEEKYELKEGQTLDVK | |
| RUO | |
| 54210 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| CL7111 | |
| Western Blot 1 μg/mL, Knockdown Validated | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction