missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TRAT1 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16881404
|
Proteintech
13279-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16871404
|
Proteintech
13279-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.Specifications
| TRAT1 | |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q3UU67, Q6PIZ9 | |
| 498075, 50852, 77647 | |
| TRAT1 Fusion Protein Ag4023 | |
| Primary | |
| -20°C | |
| TRAT1 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HSPC062, pp29/30, TCRIM, TRAT1, TRIM | |
| TRAT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title