missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13279-1-AP-150UL
This item is not returnable.
View return policy
Description
TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.Specifications
| TRAT1 | |
| Polyclonal | |
| Unconjugated | |
| TRAT1 | |
| HSPC062, pp29/30, TCRIM, TRAT1, TRIM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 498075, 50852, 77647 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UU67, Q6PIZ9 | |
| TRAT1 | |
| TRAT1 Fusion Protein Ag4023 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction