missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70735
This item is not returnable.
View return policy
Description
TRAPPC2L Polyclonal specifically detects TRAPPC2L in Human samples. It is validated for Western Blot.
Specifications
| TRAPPC2L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| TRAPPC2L | |
| Synthetic peptides corresponding to TRAPPC2L(trafficking protein particle complex 2-like) The peptide sequence was selected from the N terminal of TRAPPC2L. Peptide sequence MAVCIAVIAKENYPLYIRSTPTENELKFHYMVHTSLDVVDEKISAMGKAL. | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: 2-like protein. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| MGC111156, trafficking protein particle complex 2-like, trafficking protein particle complex subunit 2-like protein | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51693 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion