missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £386.00
Specifications
| Antigen | TRAPPC2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17762924
|
Invitrogen™
PA5121528 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17752924
![]() |
Invitrogen™
PA5121527 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is thought to be part of a large multi-subunit complex involved in the targeting and fusion of endoplasmic reticulum-to-Golgi transport vesicles with their acceptor compartment. In addition, the encoded protein can bind c-myc promoter-binding protein 1 and block its transcriptional repression capability. Mutations in this gene are a cause of spondyloepiphyseal dysplasia tarda (SEDT). A processed pseudogene of this gene is located on chromosome 19, and other pseudogenes are found on chromosomes 8 and Y. Alternatively spliced transcript variants have been found for this gene.Specifications
| TRAPPC2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P0DI81 | |
| 6399 | |
| Met1-Ser140 (Accession P0DI81), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TRAPPC2 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 1110066L09Rik; 1810064C02Rik; AW496358; hYP38334; hypothetical protein LOC616748; MBP-1-interacting protein 2A; MIP2A; MIP-2A; RGD1306925; SEDL; Sedlin; SEDLP1; SEDT; spondyloepiphyseal dysplasia tarda protein; spondyloepiphyseal dysplasia, late; trafficking protein particle complex 2; trafficking protein particle complex protein 2; Trafficking protein particle complex subunit 2; Trafficking protein particle complex subunit 2B; Trappc2; TRAPPC2.19; TRAPPC2B; TRAPPC2P1; TRS20; zgc:152903; ZNF547L | |
| TRAPPC2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title