missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121527
This item is not returnable.
View return policy
Description
The protein encoded by this gene is thought to be part of a large multi-subunit complex involved in the targeting and fusion of endoplasmic reticulum-to-Golgi transport vesicles with their acceptor compartment. In addition, the encoded protein can bind c-myc promoter-binding protein 1 and block its transcriptional repression capability. Mutations in this gene are a cause of spondyloepiphyseal dysplasia tarda (SEDT). A processed pseudogene of this gene is located on chromosome 19, and other pseudogenes are found on chromosomes 8 and Y. Alternatively spliced transcript variants have been found for this gene.
Specifications
| TRAPPC2 | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC2 | |
| 1110066L09Rik; 1810064C02Rik; AW496358; hYP38334; hypothetical protein LOC616748; MBP-1-interacting protein 2A; MIP2A; MIP-2A; RGD1306925; SEDL; Sedlin; SEDLP1; SEDT; spondyloepiphyseal dysplasia tarda protein; spondyloepiphyseal dysplasia, late; trafficking protein particle complex 2; trafficking protein particle complex protein 2; Trafficking protein particle complex subunit 2; Trafficking protein particle complex subunit 2B; Trappc2; TRAPPC2.19; TRAPPC2B; TRAPPC2P1; TRS20; zgc:152903; ZNF547L | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6399 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P0DI81 | |
| TRAPPC2 | |
| Met1-Ser140 (Accession P0DI81), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction