missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Transport Sign

Brand:  Brady™ 227795

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11760274

  • £21.80 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Plastic
Permanent Acrylic
Transport signs
297.00 mm
Toxic Gas (Flammable or Non Flammable)
Black on White
1 Ea.
Magnetic Plastic
297 mm
Diamond
Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers