missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human MyD88 siRNA
Blocks transcription and translation of MyD88 from Human.
£227.54 - £336.64
Specifications
Accession Number | NP_002459 |
---|---|
Cell Lines Tested | 293T Human Embryonic Kidney Cells with Large T Antigen. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | MYD88 |
Gene ID (Entrez) | 4615 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079191
|
enQuireBio™
QX25-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069191
|
enQuireBio™
QX25-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002459 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
4615 | |
MYD88D, Myeloid differentiation primary response protein, Toll-Receptor Signaling Cascade | |
Lyophilized |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
MYD88 | |
siRNA | |
Human |