missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human MyD88 siRNA
Blocks transcription and translation of MyD88 from Human.
Brand: enQuireBio™ QX25-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_002459 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
4615 | |
10 nmol | |
Human |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
MYD88 | |
siRNA | |
MYD88D, Myeloid differentiation primary response protein, Toll-Receptor Signaling Cascade | |
Lyophilized |