missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human IGF1R siRNA
Blocks transcription and translation of IGF1R from Human.
Brand: enQuireBio™ QX46-5nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_000866 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3480 | |
5 nmol | |
Human |
ME Human Melanoma Cells, MDA-MB-231 Human Breast Cancer Cells, and UC101 Ovarian Cancer Cells. | |
IGF1R | |
siRNA | |
JTK13, CD221, IGFIR, IGFR, Tyrosine-protein kinase receptor Insulin-like growth factor 1 receptor | |
Lyophilized |