missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human IGF1R siRNA
Blocks transcription and translation of IGF1R from Human.
£227.54 - £336.64
Specifications
Accession Number | NP_000866 |
---|---|
Cell Lines Tested | ME Human Melanoma Cells, MDA-MB-231 Human Breast Cancer Cells, and UC101 Ovarian Cancer Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | IGF1R |
Gene ID (Entrez) | 3480 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029241
|
enQuireBio™
QX46-1NMOL |
1 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039241
|
enQuireBio™
QX46-5NMOL |
5 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000866 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3480 | |
JTK13, CD221, IGFIR, IGFR, Tyrosine-protein kinase receptor Insulin-like growth factor 1 receptor | |
Lyophilized |
ME Human Melanoma Cells, MDA-MB-231 Human Breast Cancer Cells, and UC101 Ovarian Cancer Cells. | |
IGF1R | |
siRNA | |
Human |