missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ErbB3 / HER3 siRNA
Blocks transcription and translation of ErbB3 / HER3 from Human.
Brand: enQuireBio™ QX20-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_001973 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2065 | |
10 nmol | |
Human |
Kas-6/1 cells. | |
ERBB3 | |
siRNA | |
HER3, c-erbB-3, LCCS2, MDA-BF-1, c-erbB3, erbB3-S, ErbB-3, p180-ErbB3, p45-sErbB3, p85-sErbB3 | |
Lyophilized |