missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ErbB3 / HER3 siRNA
Blocks transcription and translation of ErbB3 / HER3 from Human.
£227.54 - £336.64
Specifications
Accession Number | NP_001973 |
---|---|
Cell Lines Tested | Kas-6/1 cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | ERBB3 |
Gene ID (Entrez) | 2065 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079181
|
enQuireBio™
QX20-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069181
|
enQuireBio™
QX20-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001973 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2065 | |
HER3, c-erbB-3, LCCS2, MDA-BF-1, c-erbB3, erbB3-S, ErbB-3, p180-ErbB3, p45-sErbB3, p85-sErbB3 | |
Lyophilized |
Kas-6/1 cells. | |
ERBB3 | |
siRNA | |
Human |