missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54329
This item is not returnable.
View return policy
Description
Transglutaminase 5 Polyclonal specifically detects Transglutaminase 5 in Human samples. It is validated for Western Blot.
Specifications
| Transglutaminase 5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase 5, TG(X), TGase X, TGase-5, TGASE5, TGASEX, TGM6, TGMX, TGXMGC141907, transglutaminase 5, transglutaminase V, Transglutaminase X, transglutaminase-5 | |
| Rabbit | |
| 81 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O43548 | |
| TGM5 | |
| Synthetic peptides corresponding to TGM5(transglutaminase 5) The peptide sequence was selected from the C terminal of TGM5. Peptide sequence VLKPQHQASIILETVPFKSGQRQIQANMRSNKFKDIKGYRNVYVDFAL. | |
| Affinity purified | |
| RUO | |
| 9333 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido