missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 2/TGM2 Rabbit anti-Human, Clone: 13, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | Transglutaminase 2/TGM2 |
|---|---|
| Clone | 13 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18226526
|
Novus Biologicals
NBP2-89793-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257027
|
Novus Biologicals
NBP2-89793-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transglutaminase 2/TGM2 Monoclonal specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for ELISA.Specifications
| Transglutaminase 2/TGM2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
| Recombinant Human Transglutaminase 2/TGM2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 13 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 7052 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title