missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 2/TGM2 Rabbit anti-Human, Clone: 13, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89793-100UL
This item is not returnable.
View return policy
Description
Transglutaminase 2/TGM2 Monoclonal specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for ELISA.
Specifications
| Transglutaminase 2/TGM2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 13 | |
| ELISA 1:5000-1:10000 | |
| C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
| Recombinant Human Transglutaminase 2/TGM2 protein | |
| 100 μg | |
| Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
| 7052 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction