missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Transferrin From Human Serum, Alexa Fluor™ Conjugate

Product Code. 11550756
Change view
Click to view available options
Dye Type:
Alexa Fluor Dyes
Classic Dyes
Quantity:
5 mg
Unit Size:
5mg
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dye Type Quantity Unit Size
11550756 Alexa Fluor Dyes 5 mg 5mg
11570766 Classic Dyes 5 mg 5mg
11530766 Alexa Fluor Dyes 5 mg 5mg
11590776 Alexa Fluor Dyes 5 mg 5mg
11540766 Alexa Fluor Dyes 5 mg 5mg
11560756 Alexa Fluor Dyes 5 mg Each
11510766 Alexa Fluor Dyes 5 mg 5mg
11550766 Alexa Fluor Dyes 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11550756 Supplier Invitrogen™ Supplier No. T13342
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Transferrin is a monomeric serum glycoprotein that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis—our labeled LDL complexes are useful tools for studying this phenomenon.

Fluorescent transferrin for receptor-mediated endocytosis and recycling

Fluorescent transferrin conjugates are widely used to study transferrin receptor-mediated endocytosis, endosomal trafficking, and recycling using live-cell imaging or fixed-cell endpoint analysis. Transferrin binds transferrin receptor 1 (TfR1) at the cell surface and is internalized through clathrin-mediated endocytosis. Following endosomal acidification and iron release, apo-transferrin remains bound to TfR1 and the complex recycles to the plasma membrane, where apo-transferrin dissociates.

Why use fluorescent transferrin for endocytosis studies?

  • Track receptor-mediated endocytosis: Follow transferrin binding, TfR-mediated internalization, and intracellular trafficking.
  • Monitor endocytic recycling: Use fluorescent transferrin as a well-established marker of the endosomal recycling pathway.
  • Study trafficking kinetics: Follow uptake and recycling using live-cell imaging, pulse-chase experiments, or fixed-cell endpoint analysis.
  • Assess endosomal trafficking: Investigate trafficking events associated with endosomal acidification, cargo sorting, and membrane transport.
  • Compare trafficking pathways: Combine fluorescent transferrin with fluorescent LDL or other cargo markers to compare recycling and lysosomally directed pathways.
  • Enable multiplex fluorescence imaging: Choose from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 conjugates for multicolor imaging workflows.

Fluorescent transferrin enables a direct fluorescence readout of a well-characterized endocytic pathway, making it a versatile probe for studying receptor internalization, endosomal trafficking, and recycling.

Choose a fluorescent transferrin conjugate for your imaging workflow

Select from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 transferrin conjugates, with fluorescence spanning green to far-red wavelengths.

Multiple fluorophore options

  • Match your imaging system: Select a conjugate compatible with available excitation sources, emission filters, and detectors.
  • Support multicolor imaging: Choose spectrally distinct labels for use with antibodies, organelle markers, and other fluorescent probes.
  • Optimize experimental design: Select the excitation and emission profile that best fits your available fluorescence channels.
  • Use one biological probe across workflows: Study transferrin receptor trafficking with fluorophore options suitable for a range of fluorescence microscopy applications.

Applications of fluorescent transferrin

Fluorescent transferrin is a well-established probe for studying transferrin receptor-mediated endocytosis and intracellular trafficking.

Use fluorescent transferrin to:

  • Visualize receptor-mediated internalization and recycling using live-cell imaging or fixed-cell endpoint analysis
  • Track endosomal trafficking using fluorescence and confocal microscopy
  • Study trafficking events associated with endosomal acidification and changes in endosomal function
  • Investigate transferrin receptor dynamics, including appropriately designed FRET-based studies
  • Measure transferrin receptor binding in mammalian and parasite model systems
  • Compare recycling and lysosomal trafficking using complementary fluorescent cargo markers

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Transferrin
Dye Type Alexa Fluor Dyes
Form Solid
pH 7.2
Product Line Alexa Fluor
Quantity 5 mg
Shipping Condition Room Temperature
Excitation/Emission 495/519 nm
Protein Family Transferrin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.