Learn More
trans-Zeatin-riboside
Description
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
Solubility
Soluble in water, ethanol, and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 100 mg |
| Beilstein | 5460894 |
| Solubility Information | Soluble in water,ethanol,and methanol. |
| Formula Weight | 351.36 |
| Physical Form | Powder |
| Chemical Name or Material | trans-Zeatin-riboside |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.