missing translation for 'onlineSavingsMsg'
Learn More

trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min

Product Code. 11361668
Change view
Click to view available options
Quantity:
0.5 g
10 g
Unit Size:
0.5g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361668 10 g 10g
11351668 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361668 Supplier Thermo Scientific Alfa Aesar Supplier No. 011037.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 14323-43-4 | Cl2H6N2Pd | 211.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14323-43-4
Molecular Formula Cl2H6N2Pd
Molecular Weight (g/mol) 211.38
MDL Number MFCD00011621
InChI Key WIEZTXFTOIBIOC-UHFFFAOYSA-L
Synonym diammine dichloropalladium, trans-diamminedichloropalladium ii, transpalladin, cl2pd.2nh3, trans-bisamminedichloropalladium, palladium chloride diamine, trans-dichlorodiammine palladium ii, trans-diamminedichloropalladium ii , premion
PubChem CID 61732
IUPAC Name azane;dichloropalladium
SMILES N.N.[Cl-].[Cl-].[Pd++]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 10 g
UN Number UN3077
Solubility Information Slightly soluble in water; Soluble in NH4OH; Insoluble in chloroform, acetone
Formula Weight 211.37
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 49.9% min.
Density 2.5g/mL
Assay (metals basis)
Chemical Name or Material trans-Diamminedichloropalladium(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.