accessibility menu, dialog, popup

UserName

NNClClPd

CAS RN 14323-43-4

IUPAC Name: palladium(2+) diamine dichloride
Synonyms: palladium(2+) diamine dichloride
Molecular Weight (g/mol): 211.38
Molecular Formula: Cl2H6N2Pd
InChi Key: WIEZTXFTOIBIOC-UHFFFAOYSA-L
SMILES: N.N.[Cl-].[Cl-].[Pd++]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results