missing translation for 'onlineSavingsMsg'
Learn More

trans-Clopenthixol Succinate Salt, TRC

Product Code. 30357649
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30357649 50 mg
30360837 25 mg
30359188 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357649 Supplier Toronto Research Chemicals Supplier No. TRCC58719050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246833-58-8
Molecular Formula C22 H25 Cl N2 O S . C4 H6 O4
Molecular Weight (g/mol) 519.05
Synonym 4-[3-(2-Chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol Succinate, Butanedioic acid, 2-[4-[3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol, Clopenthixol Succinate, Zuclopenthixol Succinate Salt
IUPAC Name butanedioic acid;2-[4-[3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCC=C2c3ccccc3Sc4ccc(Cl)cc24)CC1.OC(=O)CCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H25ClN2OS.C4H6O4/c23-17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)27-22)5-3-9-24-10-12-25(13-11-24)14-15-26;5-3(6)1-2-4(7)8/h1-2,4-8,16,26H,3,9-15H2;1-2H2,(H,5,6)(H,7,8)
Formula Weight 518.1642
Chemical Name or Material Clopenthixol Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.