CAS RN 1246833-58-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-Clopenthixol Succinate Salt, TRC
CAS: 1246833-58-8 Molecular Formula: C22 H25 Cl N2 O S . C4 H6 O4 Molecular Weight (g/mol): 519.05 Synonym: 4-[3-(2-Chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol Succinate,Butanedioic acid,2-[4-[3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol,Clopenthixol Succinate,Zuclopenthixol Succinate Salt IUPAC Name: butanedioic acid;2-[4-[3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol SMILES: OCCN1CCN(CCC=C2c3ccccc3Sc4ccc(Cl)cc24)CC1.OC(=O)CCC(=O)O