missing translation for 'onlineSavingsMsg'
Learn More

(-)-trans-4-[4-(4’-Fluorophenyl)-3-piperidinylmethoxy]-2-methoxyphenol(Paroxetine metabolite), TRC

Product Code. 30407908
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407908 25 mg
30397694 5 mg
30406445 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407908 Supplier Toronto Research Chemicals Supplier No. TRCF59574025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112058-90-9
Molecular Formula C19 H22 F N O3
Molecular Weight (g/mol) 331.38
Synonym 4-[[(3S,4R)-4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol, (3S-trans)-4-[[4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol, BRL 36610A
IUPAC Name 4-[[(3S,4R)-4-(4-fluorophenyl)piperidin-3-yl]methoxy]-2-methoxyphenol
SMILES COc1cc(OC[C@@H]2CNCC[C@H]2c3ccc(F)cc3)ccc1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H22FNO3/c1-23-19-10-16(6-7-18(19)22)24-12-14-11-21-9-8-17(14)13-2-4-15(20)5-3-13/h2-7,10,14,17,21-22H,8-9,11-12H2,1H3/t14-,17-/m0/s1
Formula Weight 331.1584
Chemical Name or Material (-)-trans-4-(4-(4'-Fluorophenyl)-3-piperidinylmethoxy)-2-methoxyphenol (Paroxetine metabolite)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.