CAS RN 112058-90-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-trans-4-[4-(4’-Fluorophenyl)-3-piperidinylmethoxy]-2-methoxyphenol(Paroxetine metabolite), TRC
CAS: 112058-90-9 Molecular Formula: C19 H22 F N O3 Molecular Weight (g/mol): 331.38 Synonym: 4-[[(3S,4R)-4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol,(3S-trans)-4-[[4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol,BRL 36610A IUPAC Name: 4-[[(3S,4R)-4-(4-fluorophenyl)piperidin-3-yl]methoxy]-2-methoxyphenol SMILES: COc1cc(OC[C@@H]2CNCC[C@H]2c3ccc(F)cc3)ccc1O