missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hydroxycinnamic acid, 99%

Product Code. 11441788
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441788 10 g 10g
11329479 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441788 Supplier Thermo Scientific Alfa Aesar Supplier No. A14466.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14755-02-3 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14755-02-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004390
InChI Key KKSDGJDHHZEWEP-SNAWJCMRSA-N
Synonym 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 637541
ChEBI CHEBI:32357
IUPAC Name (E)-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=CC(=O)O

Specifications

Melting Point 192°C to 196°C
Quantity 10 g
Beilstein 2690254
Formula Weight 164.16
Percent Purity 99%
Chemical Name or Material trans-3-Hydroxycinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.