missing translation for 'onlineSavingsMsg'
Learn More

trans-3'-Hydroxy Cotinine Perchlorate, TRC

Product Code. 30400054
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400054 100 mg
30393961 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400054 Supplier Toronto Research Chemicals Supplier No. TRCH924600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126874-95-1
Molecular Formula C10H12N2O2 . HClO4
Molecular Weight (g/mol) 292.67
Synonym (3R,5S)-3-Hydroxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone Perchlorate, (3R-trans)-3-Hydroxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone Monoperchlorate (Salt)
IUPAC Name (3R,5S)-3-hydroxy-1-methyl-5-pyridin-3-ylpyrrolidin-2-one;perchloric acid
SMILES CN1[C@@H](C[C@@H](O)C1=O)c2cccnc2.OCl(=O)(=O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N2O2.ClHO4/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7;2-1(3,4)5/h2-4,6,8-9,13H,5H2,1H3;(H,2,3,4,5)/t8-,9+;/m0./s1
Formula Weight 292.0462
Chemical Name or Material trans-3'-Hydroxy Cotinine Perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.