CAS RN 126874-95-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-3'-Hydroxy Cotinine Perchlorate, TRC
CAS: 126874-95-1 Molecular Formula: C10H12N2O2 . HClO4 Molecular Weight (g/mol): 292.67 Synonym: (3R,5S)-3-Hydroxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone Perchlorate,(3R-trans)-3-Hydroxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone Monoperchlorate (Salt) IUPAC Name: (3R,5S)-3-hydroxy-1-methyl-5-pyridin-3-ylpyrrolidin-2-one;perchloric acid SMILES: CN1[C@@H](C[C@@H](O)C1=O)c2cccnc2.OCl(=O)(=O)=O