missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Bromocinnamic acid, 98+%

Product Code. 11498676
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498676 5 g 5g
11408686 25 g 25g
11418686 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498676 Supplier Thermo Scientific Alfa Aesar Supplier No. A16035.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14473-91-7 | C9H7BrO2 | 227.06 g/mol

trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14473-91-7
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004382
InChI Key YEMUSDCFQUBPAL-SNAWJCMRSA-N
Synonym 3-bromocinnamic acid, 3-3-bromophenyl acrylic acid, m-bromocinnamic acid, trans-3-bromocinnamic acid, cinnamic acid, m-bromo, bw a800u, e-3-3-bromophenyl acrylic acid, 3-3-bromophenyl-2-propenoic acid, 2e-3-3-bromophenyl acrylic acid, bromocinnamic acid,3
PubChem CID 776461
IUPAC Name (E)-3-(3-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC(Br)=C1

Specifications

Melting Point 178°C to 182°C
Odor Odorless
Quantity 5 g
Beilstein 2084230
Solubility Information Insoluble in water.
Formula Weight 227.06
Percent Purity ≥98%
Chemical Name or Material trans-3-Bromocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.