Learn More
trans-3-(Benzyloxymethyl)cyclobutanol, 97%, Thermo Scientific Chemicals
Description
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 192.25 |
| Percent Purity | 97% |
| Chemical Name or Material | trans-3-(Benzyloxymethyl)cyclobutanol |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.