accessibility menu, dialog, popup

UserName

CAS RN 172324-65-1

IUPAC Name: 3-[(benzyloxy)methyl]cyclobutan-1-ol
Synonyms: 3-[(benzyloxy)methyl]cyclobutan-1-ol
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: KVQOEODTOJCIPA-UHFFFAOYSA-N
SMILES: OC1CC(COCC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results