missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hydroxycinnamic acid, 98+%

Product Code. 11479273
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479273 500 g 500g
11459273 25 g 25g
11469273 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11740.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 614-60-8 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-60-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004379
InChI Key PMOWTIHVNWZYFI-AATRIKPKSA-N
Synonym 2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
PubChem CID 637540
ChEBI CHEBI:18125
IUPAC Name (E)-3-(2-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 500 g
Beilstein 1100900
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material trans-2-Hydroxycinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.