Learn More
trans-2-Bromocinnamic acid, 98+%, Thermo Scientific Chemicals
£41.20 - £478.00
Chemical Identifiers
CAS | 7345-79-1 |
---|---|
Molecular Formula | C9H7BrO2 |
Molecular Weight (g/mol) | 227.06 |
MDL Number | MFCD00016836 |
InChI Key | OMHDOOAFLCMRFX-UHFFFAOYSA-N |
Synonym | 2-bromocinnamic acid, trans-2-bromocinnamic acid, o-bromocinnamic acid, 3-2-bromophenyl acrylic acid, bromocinnamic acid,2, 2e-3-2-bromophenyl prop-2-enoic acid, e-3-2-bromophenyl acrylic acid, 3-2-bromo-phenyl-acrylic acid, e-3-2-bromophenyl prop-2-enoic acid, 2e-3-2-bromophenyl acrylic acid |
PubChem CID | 688321 |
IUPAC Name | (E)-3-(2-bromophenyl)prop-2-enoic acid |
SMILES | OC(=O)C=CC1=CC=CC=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430203
|
Thermo Scientific Alfa Aesar
A11381.06 |
5 g |
£41.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440203
|
Thermo Scientific Alfa Aesar
A11381.14 |
25 g |
£166.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450203
|
Thermo Scientific Alfa Aesar
A11381.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7345-79-1 | |
227.06 | |
OMHDOOAFLCMRFX-UHFFFAOYSA-N | |
688321 | |
OC(=O)C=CC1=CC=CC=C1Br |
C9H7BrO2 | |
MFCD00016836 | |
2-bromocinnamic acid, trans-2-bromocinnamic acid, o-bromocinnamic acid, 3-2-bromophenyl acrylic acid, bromocinnamic acid,2, 2e-3-2-bromophenyl prop-2-enoic acid, e-3-2-bromophenyl acrylic acid, 3-2-bromo-phenyl-acrylic acid, e-3-2-bromophenyl prop-2-enoic acid, 2e-3-2-bromophenyl acrylic acid | |
(E)-3-(2-bromophenyl)prop-2-enoic acid |
Specifications
7345-79-1 | |
Odorless | |
MFCD00016836 | |
2-bromocinnamic acid, trans-2-bromocinnamic acid, o-bromocinnamic acid, 3-2-bromophenyl acrylic acid, bromocinnamic acid,2, 2e-3-2-bromophenyl prop-2-enoic acid, e-3-2-bromophenyl acrylic acid, 3-2-bromo-phenyl-acrylic acid, e-3-2-bromophenyl prop-2-enoic acid, 2e-3-2-bromophenyl acrylic acid | |
OC(=O)C=CC1=CC=CC=C1Br | |
227.06 | |
227.06 | |
trans-2-Bromocinnamic acid |
216°C to 219°C | |
C9H7BrO2 | |
2357933 | |
OMHDOOAFLCMRFX-UHFFFAOYSA-N | |
(E)-3-(2-bromophenyl)prop-2-enoic acid | |
688321 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 231-353-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only