missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, TRC

Product Code. 30378037
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378037 100 mg
30382660 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378037 Supplier Toronto Research Chemicals Supplier No. TRCD431560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61441-65-4
Molecular Formula C14 H14 Cl2 N2 O3
Molecular Weight (g/mol) 329.18
Synonym trans-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol (9CI)
IUPAC Name [(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methanol
SMILES OC[C@@H]1CO[C@@](Cn2ccnc2)(O1)c3ccc(Cl)cc3Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H14Cl2N2O3/c15-10-1-2-12(13(16)5-10)14(8-18-4-3-17-9-18)20-7-11(6-19)21-14/h1-5,9,11,19H,6-8H2/t11-,14+/m1/s1
Formula Weight 328.0381
Chemical Name or Material trans-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.