CAS RN 61441-65-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, TRC
CAS: 61441-65-4 Molecular Formula: C14 H14 Cl2 N2 O3 Molecular Weight (g/mol): 329.18 Synonym: trans-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol (9CI) IUPAC Name: [(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methanol SMILES: OC[C@@H]1CO[C@@](Cn2ccnc2)(O1)c3ccc(Cl)cc3Cl