missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanediol, 98%, Thermo Scientific Chemicals
£28.20 - £83.50
Chemical Identifiers
CAS | 1460-57-7 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00063611 |
InChI Key | PFURGBBHAOXLIO-PHDIDXHHSA-N |
Synonym | trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol |
PubChem CID | 92886 |
ChEBI | CHEBI:16931 |
IUPAC Name | (1R,2R)-cyclohexane-1,2-diol |
SMILES | O[C@@H]1CCCC[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246020
|
Thermo Scientific Acros
161650100 |
10 g | Glass bottle |
£28.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685022
|
Thermo Scientific Acros
161650500 |
50 g | Glass bottle |
£83.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
1460-57-7 | |
116.16 | |
PFURGBBHAOXLIO-PHDIDXHHSA-N | |
92886 | |
(1R,2R)-cyclohexane-1,2-diol |
C6H12O2 | |
MFCD00063611 | |
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
CHEBI:16931 | |
O[C@@H]1CCCC[C@H]1O |
Spezifikation
1460-57-7 | |
White | |
134°C | |
98% | |
C6H10(OH)2 | |
17,91 | |
Solubility in water: 375g/L (20 c) | |
O[C@@H]1CCCC[C@H]1O | |
116.16 | |
CHEBI:16931 | |
116.16 | |
Crystalline Powder or Crystals |
100°C to 104°C | |
117°C (17.0 Mbar) | |
Authentic | |
C6H12O2 | |
MFCD00063611 | |
trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
PFURGBBHAOXLIO-PHDIDXHHSA-N | |
(1R,2R)-cyclohexane-1,2-diol | |
92886 | |
8 mPa.s (110°C) | |
98% | |
trans-1, 2-Cyclohexanediol, 98% |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 215-956-6
RUO – Research Use Only