missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 97%

Product Code. 10685022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10685022 50 g Glass bottle 50g
10246020 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10685022 Supplier Thermo Scientific Acros Supplier No. 161650500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 100°C to 104°C
Color White
Boiling Point 117°C (17.0 Mbar)
Flash Point 134°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 50 g
Fieser 17,91
Solubility Information Solubility in water: 375g/L (20 c)
Viscosity 8 mPa.s (110°C)
Formula Weight 116.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-1, 2-Cyclohexanediol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.