missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM2 Antibody (SR1741), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | TRAM2 |
|---|---|
| Clone | SR1741 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651805
|
Novus Biologicals
NBP3-21638-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641174
|
Novus Biologicals
NBP3-21638-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAM2 Monoclonal antibody specifically detects TRAM2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TRAM2 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9697 | |
| IgG | |
| Affinity purified |
| SR1741 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| KIAA0057TRAM-like protein, translocating chain-associated membrane protein 2, translocation associated membrane protein 2 | |
| A synthesized peptide derived from human TRAM2 (Uniprot #: Q15035) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title