missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM2 Antibody (SR1741), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21638-50ul
This item is not returnable.
View return policy
Description
TRAM2 Monoclonal antibody specifically detects TRAM2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TRAM2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9697 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1741 | |
| Western Blot 1:500-1:2000 | |
| KIAA0057TRAM-like protein, translocating chain-associated membrane protein 2, translocation associated membrane protein 2 | |
| A synthesized peptide derived from human TRAM2 (Uniprot #: Q15035) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction