missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £234.00
Specifications
| Antigen | TRAIP |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18095994
|
Novus Biologicals
NB100-56180SS |
0.025 mL |
£157.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020146
|
Novus Biologicals
NB100-56180 |
0.05 mL |
£234.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAIP Polyclonal specifically detects TRAIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TRAIP | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| Whole antisera with 0.05% Sodium Azide | |
| 10293 | |
| A synthetic peptide corresponding to amino acids 294-313 (ESPAPVEVNLKLRRPSFRDD) of human TRIP was used as immunogen; GenBank no. gi 40807469 ref NP_005870.2 . | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein | |
| TRAIP | |
| IgG | |
| Unpurified | |
| TRIP |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title