missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56180
This item is not returnable.
View return policy
Description
TRAIP Polyclonal specifically detects TRAIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TRAIP | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TRIP | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| TRAIP | |
| A synthetic peptide corresponding to amino acids 294-313 (ESPAPVEVNLKLRRPSFRDD) of human TRIP was used as immunogen; GenBank no. gi 40807469 ref NP_005870.2 . | |
| 0.05 mL | |
| Zinc Finger | |
| 10293 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction