missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR3/TNFRSF10C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.00 - £310.00
Specifications
| Antigen | TRAILR3/TNFRSF10C |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18000094
|
Novus Biologicals
NB100-56445SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065965
|
Novus Biologicals
NB100-56445 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAILR3/TNFRSF10C Polyclonal specifically detects TRAILR3/TNFRSF10C in Human, Mouse samples. It is validated for Western Blot.Specifications
| TRAILR3/TNFRSF10C | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
| TNFRSF10C | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used is 92% homologous in DcTrailR2. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| 8794 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 155-168 ([BLAST]CSSSVSNPRNWLFL[/BLAST]) of mouse DcTrailR1; GenBank no: NP_077252.2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title