missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR3/TNFRSF10C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56445
This item is not returnable.
View return policy
Description
TRAILR3/TNFRSF10C Polyclonal specifically detects TRAILR3/TNFRSF10C in Human, Mouse samples. It is validated for Western Blot.
Specifications
| TRAILR3/TNFRSF10C | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| TNFRSF10C | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 155-168 ([BLAST]CSSSVSNPRNWLFL[/BLAST]) of mouse DcTrailR1; GenBank no: NP_077252.2. | |
| 0.1 mg | |
| Apoptosis, Breast Cancer | |
| 8794 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used is 92% homologous in DcTrailR2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction