missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £248.00
Specifications
| Antigen | TRAF-3 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030056
|
Novus Biologicals
NB100-56175SS |
0.025 mL |
£168.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004394
|
Novus Biologicals
NB100-56175 |
0.05 mL |
£248.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-3 Polyclonal specifically detects TRAF-3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| TRAF-3 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| Whole antisera with 0.02% Sodium Azide | |
| 7187 | |
| A synthetic peptide corresponding to amino acis 5-25 (KKMDSPGALQTNPPLKLHTDR) of human TRAF3 was used as immunogen; GenBank no. gi 22027616 ref NP_003291.2 . | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| TRAF3 | |
| IgG | |
| Unpurified | |
| TRAF3 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title