missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56175SS
This item is not returnable.
View return policy
Description
TRAF-3 Polyclonal specifically detects TRAF-3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TRAF-3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TRAF3 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| TRAF3 | |
| A synthetic peptide corresponding to amino acis 5-25 (KKMDSPGALQTNPPLKLHTDR) of human TRAF3 was used as immunogen; GenBank no. gi 22027616 ref NP_003291.2 . | |
| 0.025 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 7187 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction