Learn More
Gilson™ TRACKMAN Connected Starter Kit, Single Channel
The TRACKMAN Connected Starter Kit includes the pipette models that match with your QIAGEN protocols. When pairing TRACKMAN Connected with PIPETMAN™ M Connected, the installed app guides you through your QIAGEN-programmed protocol while automatically adjusting the pipette volume settings.
£3810.00
Specifications
Compatible Tips | 5 mL, 200 μL, 1200 μL tips |
---|---|
Adjustable Tip Spacing | No |
Autoclavable | Not Autoclavable |
Color | Assorted |
Dispensing Speed | Variable |
Description
We've partnered with QIAGEN, whose well-known kits help your molecular biology applications. You can now execute your QIAGEN protocols with guidance from the TRACKMAN™ Connected pipetting system. Our pipettes and tips match perfectly with the volumes needed to start your protocol, all available in one unique kit.
The TRACKMAN Connected Starter Kit single channel includes the following:
- TRACKMAN Connected tablet and accessories
- PIPETMAN M Connected P200M
- PIPETMAN™ M Connected P1200M
- PIPETMAN M Connected P5000M
- 1 rack of 5 mL tips
- 5 racks of 200 μL filter tips
- 5 racks of 1200 μL filter tips
Optimize your protocol execution with TRACKMAN Connected Starter Kit single channel and QIAGEN kits for the purification or extraction of DNA or RNA from blood, tissue, cells, virus, plasmid creation in a wide range of sample types.
- NOTE: All QIAGEN kits are sold separately and must be purchased through QIAGEN directly.
Our TRACKMAN Connected Starter Kit single channel is compatible with:
- DNeasy Blood and Tissue Mini Kit: DNA purification from blood or tissue samples.
- QIAamp DNA Mini Kit: DNA purification from human tissues, swabs, CSF, blood, body fluids, or washed cells from urine samples.
- QIAprep Spin Miniprep Kit: Plasmid DNA purification of up to 20 μg high-purity plasmid or cosmid DNA.
- QIAquick PCR Purification Kit: PCR product purification of DNA amplicons ranging from 100 bp to 10 kb.
- QIAamp ccfDNA/RNA Kit: Cell-free DNA and RNA purification from plasma and serum samples.
- RNeasy Plus Mini Kit: RNA purification from cells and tissues using gDNA Eliminator columns.
- QIAamp Viral RNA Mini Kit: Viral RNA purification from cell-free body fluids. Suitable for SARS-CoV-2 detection
Specifications
5 mL, 200 μL, 1200 μL tips | |
Not Autoclavable | |
Variable | |
All Pipettes with Tips Mounted | |
Polypropylene | |
1 | |
Yes | |
Pipette Controller | |
Pipette tips |
No | |
Assorted | |
Reusable | |
Ergonomic | |
TRACKMAN Connected | |
0.45 μm | |
Non-sterile | |
5000 μL | |
Certified Quality |