missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ TRACKMAN Connected Starter Kit, Single Channel

Product Code. 17359243 Shop All Gilson Products
Change view
Click to view available options
Plug Type:
UK-Plug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type
17359243 UK-Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17359243 Supplier Gilson™ Supplier No. FB10201
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TRACKMAN Connected Starter Kit includes the pipette models that match with your QIAGEN protocols. When pairing TRACKMAN Connected with PIPETMAN™ M Connected, the installed app guides you through your QIAGEN-programmed protocol while automatically adjusting the pipette volume settings.

We've partnered with QIAGEN, whose well-known kits help your molecular biology applications. You can now execute your QIAGEN protocols with guidance from the TRACKMAN™ Connected pipetting system. Our pipettes and tips match perfectly with the volumes needed to start your protocol, all available in one unique kit.

The TRACKMAN Connected Starter Kit single channel includes the following:

  • TRACKMAN Connected tablet and accessories
  • PIPETMAN M Connected P200M
  • PIPETMAN™ M Connected P1200M
  • PIPETMAN M Connected P5000M
  • 1 rack of 5 mL tips
  • 5 racks of 200 μL filter tips
  • 5 racks of 1200 μL filter tips

Optimize your protocol execution with TRACKMAN Connected Starter Kit single channel and QIAGEN kits for the purification or extraction of DNA or RNA from blood, tissue, cells, virus, plasmid creation in a wide range of sample types.

  • NOTE: All QIAGEN kits are sold separately and must be purchased through QIAGEN directly.

Our TRACKMAN Connected Starter Kit single channel is compatible with:

  • DNeasy Blood and Tissue Mini Kit: DNA purification from blood or tissue samples.
  • QIAamp DNA Mini Kit: DNA purification from human tissues, swabs, CSF, blood, body fluids, or washed cells from urine samples.
  • QIAprep Spin Miniprep Kit: Plasmid DNA purification of up to 20 μg high-purity plasmid or cosmid DNA.
  • QIAquick PCR Purification Kit: PCR product purification of DNA amplicons ranging from 100 bp to 10 kb.
  • QIAamp ccfDNA/RNA Kit: Cell-free DNA and RNA purification from plasma and serum samples.
  • RNeasy Plus Mini Kit: RNA purification from cells and tissues using gDNA Eliminator columns.
  • QIAamp Viral RNA Mini Kit: Viral RNA purification from cell-free body fluids. Suitable for SARS-CoV-2 detection
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips 5 mL, 200 μL, 1200 μL tips
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Assorted
Dispensing Speed Variable
Disposable Reusable
For Use With All Pipettes with Tips Mounted
Grip Type Ergonomic
Material Polypropylene
Model TRACKMAN Connected
Number of Channels 1
Pore Size 0.45 μm
Programmability Yes
Sterility Non-sterile
Type Pipette Controller
Volume (Metric) 5000 μL
For Use With (Equipment) Pipette tips
Plug Type UK-Plug
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.