Learn More
Fisherbrand™ Traceable™ Platinum Ultra-Accurate Digital Thermometer
Shop All Fisherbrand ProductsDescription
Includes
Probe, cable, Plastic carrying case, traceable certificate and 9V battery; Computer output supplied with unit
High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.
- Platinum sensor insures linear response and high accuracy across the entire range
- Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens, electronics manufacturing
- Membrane-covered keys are splash-proof and easy to operate
- HOLD allows user to freeze display for reading later
- Displays data in °F or °C
- Computer output supplied with unit
- Handle Probe: (range –200 to 500°C): Fast-response, waterproof, 4-wire platinum/stainless-steel 316 probe, 0.16-inch diameter x 7-inch stem length, overall length 11.5 inches; probe tip penetration of 0.3-inch, 6-foot cable
- Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Specifications
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Depth (English) | 0.5 in. |
| Depth (Metric) | 1.27 cm |
| Diameter (English) Probe | 0.16 in. |
| Diameter (Metric) Probe | 0.4 cm |
| For Use With (Application) | Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater |
| Height (English) | 5.5 in. |
| Height (Metric) | 13.97 cm |
| Includes | Platinum probe, cable, computer output capabilities (software package must be purchased separately), plastic carrying case, batteries and Traceable Certificate |
| Length (English) Cable | 9 ft. |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.