missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Platinum Ultra-Accurate Digital Thermometer

Product Code. 15244016 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-100.0°C to +200.0°C
-200°C to +500°C
Model:
Bullet™ Probe
Handle Probe
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Model unitSize
15244016 -100.0°C to +200.0°C Bullet™ Probe Each
15234016 -200°C to +500°C Handle Probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15244016

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Probe, cable, Plastic carrying case, traceable certificate and 9V battery; Computer output supplied with unit

High-precision unit is designed for years of reliable service—even in the severest environments.

High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.

  • Platinum sensor insures linear response and high accuracy across the entire range
  • Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens, electronics manufacturing
  • Membrane-covered keys are splash-proof and easy to operate
  • HOLD allows user to freeze display for reading later
  • Displays data in °F or °C
  • Computer output supplied with unit
  • Handle Probe: (range –200 to 500°C): Fast-response, waterproof, 4-wire platinum/stainless-steel 316 probe, 0.16-inch diameter x 7-inch stem length, overall length 11.5 inches; probe tip penetration of 0.3-inch, 6-foot cable
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Depth (English) 0.5 in.
Depth (Metric) 1.27 cm
Diameter (English) Probe 0.16 in.
Diameter (Metric) Probe 0.4 cm
For Use With (Application) Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater
Height (English) 5.5 in.
Height (Metric) 13.97 cm
Includes Platinum probe, cable, computer output capabilities (software package must be purchased separately), plastic carrying case, batteries and Traceable Certificate
Length (English) Cable 9 ft.
Length (English) Probe 5.875 in.
Length (Metric) 292 mm
Length (Metric) Cable 304.8 cm
Length (Metric) Probe 2.54 cm
Material Platinum, Stainless Steel
Model Bullet™ Probe
Probe Type Bullet probe
Resolution 0.0001°C
Weight (English) 8 oz.
Weight (Metric) 226.7 g
Width (English) 3.5 in.
Product Type Digital Thermometer
Length (English) 11.5 in.
Max. Temperature (English) 392°F
Max. Temperature (Metric) 200°C
Min. Temperature (English) -148°F
Min. Temperature (Metric) -100°C
Temperature Range (Metric) -100.0°C to +200.0°C
Calibrated Yes
Accuracy ±0.05°C
Display Digital LCD
Width (Metric) 8.89 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.